missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTAL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534717
This item is not returnable.
View return policy
Description
Recommended positive controls: H1299, Raji. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NTAL (Non-T cell Activation Linker), also known as LAB (Linker for activation of B cells), is a 30 kDa double-palmitoylated transmembrane adaptor protein expressed by B cells, NK cells, mast cells and macrophages. It is a negative regulator of early stages of BCR-dependent B cell signaling and serves as a negative regulator also in mast cells. However, in mast cells, NTAL also contributes to some activation processes, partially overlapping with LAT function.
Specifications
| NTAL | |
| Polyclonal | |
| Unconjugated | |
| LAT2 | |
| AW125574; HSPC046; LAB; Lat2; LAT-2 like; Linker for activation of B-cells; linker for activation of T cells family, member 2; linker for activation of T cells, transmembrane adaptor 2; linker for activation of T-cells family member 2; Membrane-associated adapter molecule; non-T cell activation linker; non-T-cell activation linker; NTAL; WBS15; WBSCR15; WBSCR5; Williams-Beuren syndrome chromosomal region 15 protein; williams-Beuren syndrome chromosomal region 15 protein homolog; Williams-Beuren syndrome chromosomal region 5 protein; Williams-Beuren syndrome chromosome region 5 homolog; Williams-Beuren syndrome chromosome region 5-like protein; WSCR5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7462 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.62 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9GZY6 | |
| LAT2 | |
| Synthetic peptide encompassing a sequence within the Intracellular domain of human NTAL. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction