missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTAN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 183.00 - € 322.00
Specifications
| Antigen | NTAN1 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18693107
|
Novus Biologicals
NBP3-12564-100ul |
100 μg |
€ 322.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619266
|
Novus Biologicals
NBP3-12564-50ul |
50 μg |
€ 183.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NTAN1 Polyclonal antibody specifically detects NTAN1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| NTAN1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS, pH7.0 | |
| 123803 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp666E058, EC 3.5.1.-, N-terminal asparagine amidase, N-terminal asparagine amidohydrolase, PNAA, PNAD, Protein NH2-terminal asparagine amidohydrolase, Protein NH2-terminal asparagine deamidase, Protein N-terminal Asn amidase, protein N-terminal asparagine amidohydrolase, Protein NTN-amidase | |
| Produced in rabbits immunized with E. coli-derived Human NTAN1 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title