Learn More
Novus Biologicals™ NTB-A/SLAMF6/CD352 Antibody (SGN-CD352A) - Humanized, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28645-1mg
 View more versions of this product
Product Code. 30047026
  • € 920.00 / 1mg
Description
NTB-A/SLAMF6/CD352 Monoclonal antibody specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NTB-A/SLAMF6/CD352
Monoclonal
Unconjugated
Q96DU3
Mouse
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
SGN-CD352A
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6
SLAMF6/ CD352
1 mg
Immunology
114836