Learn More
PeproTech NTF4 Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P83-50UG
Description
AA Sequence of recombinant protein (monomer): GVSETAPASR RGELAVCDAV SGWVTDRRTA VDLRGREVEV LGEVPAAGGS PLRQYFFETR CKADNAEEGG PGAGGGGCRG VDRRHWVSEC KAKQSYVRAL TADAQGRVGW RWIRIDTACV CTLLSRTGRA Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human NT-4. Anti-Human NT-4-specific antibody was purified by affinity chromatography employing an immobilized Human NT-4 matrix. Sandwich ELISA: To detect Human NT-4 by sandwich ELISA (using 100 μL/well) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human NT-4 (500-P83BT) as a detection antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human NT-4. Western Blot: To detect Human NT-4 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human NT-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
This gene is a member of a family of neurotrophic factors, neurotrophins, that control survival and differentiation of mammalian neurons. The expression of this gene is ubiquitous and less influenced by environmental signals. While knock-outs of other neurotrophins including nerve growth factor, brain-derived neurotrophic factor, and neurotrophin 3 prove lethal during early postnatal development, NTF5-deficient mice only show minor cellular deficits and develop normally to adulthood.
Specifications
| NTF4 | |
| Polyclonal | |
| PBS with no preservative | |
| P34130 | |
| NTF4 | |
| E.coli-derived, 28.1kDa Recombinant Human NT-4. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| Unconjugated | |
| NTF4 | |
| 2900040K06Rik; AI462899; GLC10; GLC1O; neurotrophic factor 4; neurotrophic factor 5; neurotrophin 4; Neurotrophin 4/5; neurotrophin 5; Neurotrophin 5 (neurotrophin 4/5); neurotrophin 5 (neurotrophin 4/5); neurotrophin-4; Neurotrophin-5; neutrophic factor 4; Neutrophic factor 5; NT4; NT-4; NT4/5; NT-4/5; NT4/NT5; NT4P; NT5; NT-5; Ntf4; Ntf5; Ntf-5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4909 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.