missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTH1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-108AF700
This item is not returnable.
View return policy
Description
NTH1 Polyclonal antibody specifically detects NTH1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| NTH1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| EC 4.2.99.18, endonuclease III-like protein 1, nth endonuclease III-like 1 (E. coli), OCTS3NTH1nth (E.coli endonuclease III)-like 1 | |
| A peptide derived from human NTH1, conjugated to KLH. [UniProt# P78549] | |
| 0.2 mL | |
| Base Excision Repair, Cancer, Chromatin Research, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 4913 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction