missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTH1 Antibody, DyLight 755 , Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-108IR
This item is not returnable.
View return policy
Description
NTH1 Polyclonal antibody specifically detects NTH1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| NTH1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| EC 4.2.99.18, endonuclease III-like protein 1, nth endonuclease III-like 1 (E. coli), OCTS3NTH1nth (E.coli endonuclease III)-like 1 | |
| A peptide derived from human NTH1, conjugated to KLH. [UniProt# P78549] | |
| 0.2 mL | |
| Base Excision Repair, Cancer, Chromatin Research, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 4913 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction