missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTHL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112631
This item is not returnable.
View return policy
Description
NTHL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Oxygen radicals damage chromosonal DNA causing cell death and inducing mutations. Although a major focus of oxidatively damaged DNA has centered on the repair of 8-oxo-G, a number of other damaged DNA sites are created by free-radical attack on DNA. The human NTH1 repair protein is one enzyme that has been shown to act on a large number of these other oxidatively damaged DNA sites.A homologue of E. coli Endonuclease III, NTH is a DNA glycosylase with apurinic/apyrimidinic lyase activity. The NTH protein has broad substrate specificity, including numerous ring saturation and fragmentation products of pyrimidines. Research indicates NTH plays a crucial role in removal of oxidative base lesions in mitochondrial DNA.
Specifications
| NTHL1 | |
| Polyclonal | |
| Unconjugated | |
| NTHL1 | |
| bifunctional DNA N-glycoslyase/DNA-(apurinic or apyrimidinic site) lyase; Bifunctional DNA N-glycosylase/DNA-(apurinic or apyrimidinic site) lyase; DNA glycoslyase/AP lyase; DNA glycosylase/AP lyase; Endonuclease III-like protein 1; FAP3; hNTH1; nth (endonuclease III)-like 1 (E.coli); nth endonuclease III-like 1; Nth1; NTHL 1; Nthl1; nth-like DNA glycosylase 1; OCTS 3; OCTS3; thymine glycol DNA glycosylase/AP lyase | |
| Rabbit | |
| Protein G | |
| RUO | |
| 18207, 4913 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4.61 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O35980, P78549 | |
| NTHL1 | |
| Recombinant Human Endonuclease III-like protein 1 protein (31-312AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction