missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTMT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111469
This item is not returnable.
View return policy
Description
Immunogen sequence: MTSEVIEDEK QFYSKAKTYW KQIPPTVDGM LGGYGHISSI DINSSRKFLQ RFLREGPNKT Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.
The METTL11A gene encodes an N-terminal methyltransferase for the RAN (MIM 601179) guanine nucleotide exchange factor regulator of chromosome condensation 1 (RCC1; MIM 179710). METTL11A enzyme alpha-N-methylates other protein targets such as SET (MIM 600960) and RB (MIM 180200).
Specifications
| NTMT1 | |
| Polyclonal | |
| Unconjugated | |
| NTMT1 | |
| 2610205E22Rik; AD-003; AL033331; AL033332; Alpha N-terminal protein methyltransferase 1A; C9orf32; HOMT1A; methyltransferase like 11A; methyltransferase-like protein 11A; METTL11A; NRMT; NRMT1; N-terminal RCC1 methyltransferase; N-terminal Xaa-Pro-Lys N-methyltransferase 1; N-terminal Xaa-Pro-Lys N-methyltransferase 1, N-terminally processed; NTM1A; NTMT1; RGD1306582; X-Pro-Lys N-terminal protein methyltransferase 1A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 28989 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BV86 | |
| NTMT1 | |
| Recombinant Protein corresonding to Human NTMT1. Recombinant protein control fragment (Product #RP-106854). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction