missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTN5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571162
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 92%; Guinea Pig: 92%; Human: 100%; Mouse: 77%; Pig: 100%; Rat: 77%.
NTN5 gene ontology annotations related to this gene include neurogenesis.
Specifications
| NTN5 | |
| Polyclonal | |
| Unconjugated | |
| Ntn5 | |
| Gm484; netrin 5; netrin-1-like; netrin-1-like protein; Netrin-5; NTN5; RGD1565197; zmp:0000000590 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 126147 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WTR8 | |
| Ntn5 | |
| synthetic peptide directed towards the C-terminal region of human NTN5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction