missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTNG1 Antibody (RM0292-4K62), Alexa Fluor™ 647, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11810AF647
This item is not returnable.
View return policy
Description
NTNG1 Monoclonal specifically detects NTNG1 in Mouse samples. It is validated for Western Blot.
Specifications
| NTNG1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NTNG1 | |
| Mouse recombinant NTNG1 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0292-4K62 | |
| Western Blot | |
| axon guidance molecule, KIAA0976YLSR571, laminet 1, laminet-1, Lmnt1, netrin G1, netrin G1f, netrin-G1 | |
| Rat | |
| Protein A or G purified | |
| Neuroscience | |
| 22854 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction