missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTNG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566811
This item is not returnable.
View return policy
Description
Immunogen sequence: PYPLVWGHYDL CKTQIYTEEG KVWDYMACQP ESTDMTKYLK VKLDPPDITC GD Highest antigen sequence identity to the following orthologs - mouse 89%, rat 49%.
Netrin G1 (NTNG1) belongs to a conserved family of proteins that act as axon guidance cues during vertebrate nervous system development (Nakashiba et al., 2000).
Specifications
| NTNG1 | |
| Polyclonal | |
| Unconjugated | |
| NTNG1 | |
| A930010C08Rik; AI853992; axon guidance molecule; Kiaa0976; laminet 1; laminet-1; LMNT1; netrin G1; netrin G1f; netrin-G1; Ntng1; RGD1563465; UNQ571/PRO1133; YLSR571 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22854 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2I2 | |
| NTNG1 | |
| Recombinant Human NTNG1. Recombinant protein control fragment (Product #RP-107469). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction