missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NTNG1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17187973
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17187973 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17187973 Supplier Invitrogen™ Supplier No. PA5110046

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: HYDLCKTQIY TEEGKVWDYM ACQPESTDMT KYLKVKLDPP DITCGDPPET FCAMGNPYMC NNECDASTPE LAHPPELMFD FEGRHPSTFW QSATWKEYPK PLQVNITLSW SKTIELTDNI VITFESGRPD QMILEKSLDY GRTWQPYQYY ATDCLDAFHM DPKSVKDLSQ HTVLEIICTE EYSTGYTTNS KIIHFEIKDR FAFFAGPRLR NMASLYGQLD TTKKLRDFFT VTDLRIRLLR PAVGEIFVDE LHLARYFYAI SDIKVRGRCK CN.

Netrin G1 (NTNG1) belongs to a conserved family of proteins that act as axon guidance cues during vertebrate nervous system development (Nakashiba et al., 2000).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTNG1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NTNG1
Gene Accession No. Q8R4G0, Q9Y2I2
Gene Alias A930010C08Rik; AI853992; axon guidance molecule; Kiaa0976; laminet 1; laminet-1; LMNT1; netrin G1; netrin G1f; netrin-G1; Ntng1; RGD1563465; UNQ571/PRO1133; YLSR571
Gene Symbols NTNG1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 29-300 of human NTNG1 (NP_055732.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22854, 295382, 80883
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.