missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTPCR Antibody (2C3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00084284-M04
This item is not returnable.
View return policy
Description
NTPCR Monoclonal antibody specifically detects NTPCR in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| NTPCR | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| C1orf57, chromosome 1 open reading frame 57, EC 3.6.1.15, FLJ11383, HCR-NTPase, human cancer-related NTPase, MGC13186, NTPase, Nucleoside triphosphate phosphohydrolase, nucleoside-triphosphatase C1orf57, nucleoside-triphosphatase, cancer-related, RP4-678E16.2 | |
| C1orf57 (NP_115700, 91 a.a. ∽ 190 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LALPVLRNADCSSGPGQRVCVIDEIGKMELFSQLFIQAVRQTLSTPGTIILGTIPVPKGKPLALVEEIRNRKDVKVFNVTKENRNHLLPDIVTCVQSSRK | |
| 0.1 mg | |
| Protein Kinase | |
| 84284 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 2C3 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_115700 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction