missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTRK3 Mouse anti-Human, Clone: OTI2B8, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500537S
This item is not returnable.
View return policy
Description
NTRK3 Monoclonal antibody specifically detects NTRK3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| NTRK3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q16288 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4916 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B8 | |
| Unconjugated | |
| Ntrk3 | |
| AW125844; ETS related protein-neurotrophic receptor tyrosine kinase fusion protein; ETV6-NTRK3 fusion; gp145(trkC); GP145-TrkC; neural receptor protein-tyrosine kinase (trkC); neurotrophic receptor tyrosine kinase 3; Neurotrophic tyrosine kinase receptor type 3; neurotrophic tyrosine kinase, receptor, type 3; neurotrophin-3; NT-3 growth factor receptor; NTRK3; Ntrk3_tv3; TrkC; trk-C; trkC tyrosine kinase; tyrosine kinase receptor; tyrosine kinase receptor C | |
| Full-length protein expressed in 293T cell transfected with human NTRK3 expression vector | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction