missing translation for 'onlineSavingsMsg'
Learn More

NU 7026, MedChemExpress

Product Code. p-200097741
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442847 50 mg 50mg
16439317 5 mg 5mg
16436768 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16442847 Supplier Medchem Express Supplier No. HY15719

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NU 7026 (LY293646) is a novel specific DNA-PK inhibitor with IC50 of 0.23 μM, also inhibits PI3K with IC50 of 13 μM.

NU 7026 (LY293646) is a novel specific DNA-PK inhibitor with IC50 of 0.23 μM, also inhibits PI3K with IC50 of 13 μM.

Specifications

Chemical Name or Material NU 7026
CAS 154447-35-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H15NO3
Quantity 50 mg
Synonym LY293646
Solubility Information DMSO : 2.5 mg/mL (8.89 mM; Need ultrasonic)
SMILES O=C1C2=CC=C3C=CC=CC3=C2OC(N4CCOCC4)=C1
Molecular Weight (g/mol) 281.31
Formula Weight 281.31
Percent Purity 99.43%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.