Learn More
NUB1 Mouse anti-Canine, Human, Rat, Clone: OTI4H2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF800188
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
NUB1 interacts with and negatively regulates NEDD8, a ubiquitin-like protein that covalently conjugates to cullin family members.Specifications
| NUB1 | |
| Monoclonal | |
| Unconjugated | |
| NUB1 | |
| BS4, NUB1L, NYREN18 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 296731, 475542, 51667 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4H2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9Y5A7 | |
| NUB1 | |
| Human recombit protein fragment corresponding to amino acids 1-326 of human NUB1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.