missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUBP2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94547-0.02ml
This item is not returnable.
View return policy
Description
NUBP2 Polyclonal antibody specifically detects NUBP2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NUBP2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CFD1C447E6.1 (nucleotide binding protein 1 (E.coli MinD like) ), cytosolic Fe-S cluster assembly factor NUBP2, homolog of yeast cytosolic Fe-S cluster deficient 1, NBP 2, NUBP1, nucleotide binding protein 2 (E.coli MinD like), nucleotide binding protein 2 (MinD homolog, E. coli), Nucleotide-binding protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 142-271 of human NUBP2 (NP_036357.1). MATIEALRPYQPLGALVVTTPQAVSVGDVRRELTFCRKTGLRVMGIVENMSGFTCPHCTECTSVFSRGGGEELAQLAGVPFLGSVPLDPALMRTLEEGHDFIQEFPGSPAFAALTSIAQKILDATPACLP | |
| 0.02 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology, Signal Transduction | |
| 10101 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction