Learn More
NUBPL Monoclonal Antibody (OTI3D11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503745
This item is not returnable.
View return policy
Description
Required for the assembly of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I). May deliver of one or more Fe-S clusters to complex I subunits.Specifications
| NUBPL | |
| Monoclonal | |
| 0.6 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TB37 | |
| NUBPL | |
| Human recombit protein fragment corresponding to amino acids 1-250 of human NUBPL produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D11 | |
| Unconjugated | |
| NUBPL | |
| C14orf127, IND1, huInd1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 80224 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |