Learn More
NUBPL Monoclonal Antibody (OTI3D11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA503745
Encompass_Preferred
 View more versions of this product
Product Code. 17529941
  • € 540.87 / 100µL
Description
Required for the assembly of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I). May deliver of one or more Fe-S clusters to complex I subunits.
Specifications
Specifications
NUBPL
Monoclonal
0.6 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8TB37
NUBPL
Human recombit protein fragment corresponding to amino acids 1-250 of human NUBPL produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI3D11
Unconjugated
NUBPL
C14orf127, IND1, huInd1
Mouse
Affinity Chromatography
RUO
80224
-20° C, Avoid Freeze/Thaw Cycles