Learn More
NUBPL Monoclonal Antibody (OTI3D11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503745S
This item is not returnable.
View return policy
Description
NUBPL Monoclonal antibody specifically detects NUBPL in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| NUBPL | |
| Monoclonal | |
| 0.6 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TB37 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 80224 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D11 | |
| Unconjugated | |
| Nubpl | |
| 2410170E07Rik; C14orf127; huInd1; IND1; IND1 homolog; Iron-sulfur protein NUBPL; iron-sulfur protein required for NADH dehydrogenase; NUBPL; nucleotide binding protein like; nucleotide binding protein-like; nucleotide-binding protein-like; RGD1307232 | |
| Human recombinant protein fragment corresponding to amino acids 1-250 of human NUBPL produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |