Learn More
Nuclear Antigen Antibody (235-1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-34525PE-100ul
 View more versions of this product
Product Code. 18450812
  • € 643.00 / 100µL
Description
Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Porcine, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Flow (Intracellular), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nuclear Antigen
Monoclonal
PE
PBS
Mouse
Protein A or G purified
RUO
Human, Pig, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative)
Purified
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Flow Cytometry (Intracellular Staining), CyTOF
235-1
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready
Nuclei, Nucleus
Nuclei of human myeloid leukemia biopsy cells
100 μL
Primary
Store at 4C in the dark.
IgG1 κ