missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Antigen Antibody (NM2984R), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08513FR
This item is not returnable.
View return policy
Description
Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Nuclear Antigen | |
| Monoclonal | |
| DyLight 680 | |
| Nuclei, Nucleus | |
| Nuclei of HL60 cells | |
| 100 μg | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| NM2984R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction