missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Antigen Antibody (rNM106) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14097JF585
This item is not returnable.
View return policy
Description
Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Nuclear Antigen | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Nuclei, Nucleus | |
| Nuclei of HL60 cells | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| rNM106 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction