missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Factor Erythroid 2 Related Factor 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15045
This item is not returnable.
View return policy
Description
Nuclear Factor Erythroid 2 Related Factor 1 Polyclonal specifically detects Nuclear Factor Erythroid 2 Related Factor 1 in Human samples. It is validated for Western Blot.
Specifications
| Nuclear Factor Erythroid 2 Related Factor 1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| FLJ00380, HBZ17, LCR-F1, Locus control region-factor 1, NF-E2-related factor 1, NFE2-related factor 1, NRF1TCF11, nuclear factor (erythroid-derived 2)-like 1, nuclear factor erythroid 2-related factor 1, Nuclear factor, erythroid derived 2, like 1, TCF-11, Transcription factor 11, transcription factor 11 (basic leucine zipper type), Transcription factor HBZ17, Transcription factor LCR-F1 | |
| Rabbit | |
| 85 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.51mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q14494 | |
| NFE2L1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human Nrf1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 4779 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction