missing translation for 'onlineSavingsMsg'
Learn More

Nucleobindin 1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-97441AF700

Product Code. 18070719

  • € 415.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Nucleobindin 1 Polyclonal specifically detects Nucleobindin 1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nucleobindin 1
Polyclonal
50 mM Sodium Borate
Rabbit
Antigen Affinity-purified
RUO
4924
Store at 4°C in the dark.
IgG
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 700
DKFZp686A15286, FLJ40471, NUCCALNUC, nucleobindin 1, nucleobindin-1
Produced in rabbits immunized with E. coli-derived Human Nucleobindin 1 fragment.
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.