missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleobindin 1 Antibody (OTI1A5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73086H
This item is not returnable.
View return policy
Description
Nucleobindin 1 Monoclonal specifically detects Nucleobindin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Nucleobindin 1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| NUCB1 | |
| Full length recombinant protein of human Nucleobindin 1 (NP_006175) produced in HEK293T cells. | |
| 0.1 mL | |
| 4924.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1A5 | |
| Western Blot, Immunohistochemistry | |
| DKFZp686A15286, FLJ40471, NUCCALNUC, nucleobindin 1, nucleobindin-1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction