Learn More
Macherey-Nagel™ NucleoBond™ Finalizer Large Plus for desalting and concentration of plasmid DNA
Product Code. 12765194
Click to view available options
:
No. of Reactions: 20
Product Code. 12765194
Brand: Macherey-Nagel™ 740419.20
Proven syringe filter for speeding up anion exchange plasmid preparations.
The NucleoBond Finalizers are designed for quick concentration and desalination of plasmid and cosmid DNA eluates that are obtained by anion-exchange chromatographic DNA purification with NucleoBond columns. The sample is precipitated with isopropanol and loaded onto a NucleoBond Finalizer membrane by means of a syringe. After an ethanolic washing step and drying of the membrane the pure DNA can be eluted with low salt buffer for further use. The NucleoBond Finalizer technology replaces the tedious and time-consuming centrifugation step which hitherto follows every isopropanol precipitation of diluted DNA samples. Furthermore problems like lost DNA pellets or incomplete solubilisation of hardly visible precipitates can easily be circumvented. The pure concentrated and almost salt free DNA can be used directly for all common down-stream applications like transfection, in vitro transcription, automated or manual sequencing, cloning, hybridisation, and PCR*.
  • Elimination of centrifugation steps for precipitation, time saving from >1 hr. to only 5min
  • Two sizes available, to be combined with Midi and Maxi preparations, higher flexibility
  • Highly concentrated plasmid DNA depending on elution volume
  • No loss of DNA pellets or imcomplete solubilisation of hardly visible precipitates Quick concentration and desalination of plasmid and cosmid DNA elutes obtained by anion-exchange chromatography. DNA suitable for e.g., transfection, in vitro transcription, automated or manual sequencing, cloning, hybridisation and PCR*.
Specifications
Format Syringe Filter
Isolation Technology Anion Exchange Chromatography Technology
Final Product Type Nucleo Bond Finalizer Large Plus
No. of Reactions 20
Product Type Nucleo Bond Finalizer Large Plus
Size 2-50kbp
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoBond Xtra Maxi sample
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Respiratory sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol, potassium chloride, sodiumhydroxide, dodecyl sulphate, sodium salt, RNase, ethanol