missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond RS 10 Column - Purification of Vaccine-Grade Plasmid DNA

Product Code. 18051091
Click to view available options
Quantity:
5 Preps
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 18051091

Brand: Macherey-Nagel™ 743502

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Column for preparative scale isolation of vaccine-grade plasmid DNA with peristaltic pump or FPLC system.

  • Plasmid DNA

Specifications

Cell Type Bacteria, E. coli
Capacity Up to 10 mg
Content And Storage 5 Preps
Description Preparative scale
Isolation Technology Anion Exchange Chromatography Technology
Sample Type Bacteria, E. coli
Final Product Type Plasmid DNA
For Use With (Application) Vaccine-grade plasmid DNA isolation
No. of Reactions 5 Preps
Product Line NucleoBond
Product Type Column
Quantity 5 Preps
Purity 1.8 to 2.0
Sample Volume Maximum 8 g wet weight
Technique Peristaltic pump, FPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.