missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleocapsid Antibody (1051436), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB11289PEATT594
This item is not returnable.
View return policy
Description
Nucleocapsid Monoclonal antibody specifically detects Nucleocapsid in HCoV-OC43 samples. It is validated for Flow Cytometry
Specifications
| Nucleocapsid | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Sf-21 (baculovirus)-derived hcov-oc43 Nucleocapsid, Met1-Ile448, Accession # YP_009555245.1 | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry | |
| 1051436 | |
| Flow Cytometry | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| HCoV-OC43 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction