Learn More
Nucleocapsid protein, Mouse anti-SARS-CoV-2, Clone: AS47, ACROBiosystems
HEK293 Monoclonal Antibody
Brand: ACROBiosystems NUN-S47-10mg
Description
This product is a specific antibody against SARS-CoV-2 Nucleocapsid protein. No cross-reactivity is detected with nucleocapsid protein of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-L63, HCoV-OC43 and HCoV-HKU1.(Trehalose free)Specifications
| Nucleocapsid protein | |
| AS47 | |
| Trehalose free | |
| 10 mg | |
| Primary | |
| SARS-CoV-2 | |
| Recombinant Antibodies | |
| Mouse IgG1 κ |
| ELISA | |
| Unconjugated | |
| HEK293 | |
| RUO | |
| This product is a specific antibody against SARS-CoV-2 Nucleocapsid protein. No cross-reactivity is detected with nucleocapsid protein of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-L63, HCoV-OC43 and HCoV-HKU1. | |
| 2-8°C | |
| Liquid |