Learn More
Macherey-Nagel™ Nucleodur™ C18 ec HPLC CC Guard Columns
Product Code. 12468557 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
3 mm
4 mm
Carbon Load:
17.5%
4%
Unit Size:
Each
Product Code. 12468557
Brand: Macherey-Nagel™ 761005.30
Only null left
Specifications
Carbon Load 17.5%
pH 1 to 9
Particle Size 3 μm
Length (Metric) 8 mm
Pore Size 100Å
Type CC Guard Column
Diameter (Metric) Inner 3 mm
Stationary Phase NUCLEODUR™ C18 ec
Mobile Phase Compatibility Acetonitrile/Water
Solvents Acetonitrile/Water
Packing Material Silica
Crosslinking Octadecyl
Column Format Guard Column
For Use With (Application) Scaling up of preparative HPLC
Technique HPLC
Particle Shape Spherical
Endcapped Yes
USP Type L1