Learn More
Macherey-Nagel™ Nucleodur™ HILIC HPLC Column
Product Code. p-8012230 Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
30 mm
50 mm
75 mm
100 mm
125 mm
150 mm
250 mm
Diameter (Metric):
2 mm
3 mm
4 mm
4.6 mm
Particle Size:
1.8 μm
5 μm
Unit Size:
Each
Product Code. 12728872
Brand: Macherey-Nagel™ 760551.20
Column Format: Analytical Column; Carbon Load: 7%; Pore Size: 110 Å; Column Phase: HILIC
Specifications
Column Chemistry Silica
Column Format Analytical Column
Crosslinking Ammonium
Diameter (Metric) 2 mm
Flow Rate 1 mL/min
For Use With For HPLC, GC, TLC, SPE
Functionality/Interaction Type Hydrophilic
Length (Metric) 125 mm
Max. Pressure 8700 psi (600 bar)
Mobile Phase Compatibility 80:20 Acetonitrile/Water
Particle Shape Spherical
Particle Size 5 μm
pH 2 to 8.5
Pore Size 110 Å
Solvents HILIC
Surface Area 340 m²/g
Temperature 60°C
Stationary Phase Ammonium/Sulfonic Acid Modified Silica
Packing Material Silica
Carbon Load 7%
Column Type HILIC
Endcapped No
Phase HILIC
Material Stainless Steel
Type EC Column
Technique Analytical HPLC