Learn More
Macherey-Nagel™ Nucleodur™ HILIC HPLC EC Guard Columns
Product Code. p-8012231 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
2 mm
3 mm
Unit Size:
Each
Product Code. 12708992
Brand: Macherey-Nagel™ 761960.20
Only null left
Specifications
Carbon Load 7%
pH 2 to 8.5
Particle Size 1.8 μm
Length (Metric) 4 mm
Pore Size 110Å
Type EC Guard Column
Diameter (Metric) Inner 2 mm
Stationary Phase NUCLEODUR™ HILIC
Mobile Phase Compatibility Acetonitrile/Water
Solvents Acetonitrile/Water
Packing Material Silica
Crosslinking Ammonium/Sulfonic Acid
Column Format Guard Column
For Use With (Application) Analytical and preparative applications as well as LC/MS
Technique HPLC
Particle Shape Spherical