Learn More
Macherey-Nagel™ NUCLEODUR™ HILIC Column for HPLC, 3μm Particles
Product Code. p-8012232 Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
50 mm
100 mm
125 mm
150 mm
250 mm
Diameter (Metric):
2 mm
3 mm
4 mm
4.6 mm
Flow Rate:
0.15 mL/min
0.34 mL/min
0.6 mL/min
0.8 mL/min
Unit Size:
Each
Product Code. 10668614
Brand: Macherey-Nagel™ 760534.20
Ideal for analytical HPLC applications with hydrophilic compounds. Macherey-Nagel™ NUCLEODUR™ HILIC Column for HPLC, 3μm Particles is an EC standard column with high purity silica phase. The columns are available in a variety of diameters and lengths.
This product is suitable for Hydrophilic Interaction Chromatography (HILIC) applications. It features a highly pure ammonium/sulfonic acid-modified silica phase with an 80:20 acetonitrile-water eluent. These EC standard columns have a particle size of 3μm and are used in analytical HPLC.
  • Ideal for reproducible and stable chromatography of highly polar analytes
  • Suitable for analytical and preparative applications
  • Very short column conditioning period
  • Used with hydrophilic compounds, such as: organic polar acids and bases, polar natural compounds, nucleosides, oligonucleotides, amino acids, peptides, water soluble vitamins
Specifications
Column Chemistry Ammonium /sulfonic acid
Column Format Analytical Column
Crosslinking Ammonium /sulfonic acid
Diameter (Metric) 2 mm
Flow Rate 0.15 mL/min
For Use With For Reproducible and Stable Chromatography of Highly Polar Analytes
Functionality/Interaction Type Hydrophobic
Hydrophobicity Yes
Length (Metric) 100 mm
Max. Pressure 8700 psi (600 bar)
Mobile Phase Compatibility 80:20 Acetonitrile/water
Particle Shape Spherical
Particle Size 3 μm
pH 2 to 8.5
Pore Size 110 Å
Pore Volume 0.9 mL/g
Quantity 1 Ea.
Solvents 80:20 Acetonitrile/water
Surface Area 340 m²/g
Temperature 60°C
Stationary Phase Ammonium/Sulfonic Acid Modified Silica
Packing Material Silica
Carbon Load 7%
Column Type HILIC
Endcapped No
Phase HILIC
Material Stainless Steel
Type EC Column