Learn More
Macherey-Nagel™ Nucleodur™ NH2-RP HPLC EC Guard Columns
Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
2 mm
3 mm
Unit Size:
Each
Specifications
Specifications
| Carbon Load | 2.5% |
| Particle Size | 3 μm |
| Length (Metric) | 4 mm |
| Pore Size | 100Å |
| Type | EC Guard Column |
| Diameter (Metric) Inner | 2 mm |
| Stationary Phase | NUCLEODUR™ NH2-RP |
| Mobile Phase Compatibility | Acetonitrile/Water |
| Solvents | Acetonitrile/Water |
| Packing Material | Silica |
| Show More |