Learn More
Macherey-Nagel™ Nucleodur™ PFP HPLC Columns
Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric):
2 mm
3 mm
4.6 mm
Unit Size:
Each
Specifications
Specifications
| Column Chemistry | Pentafluorophenyl-propyl |
| Column Format | Analytical Column |
| Crosslinking | Pentafluorophenyl-propyl |
| Diameter (Metric) | 4.6 mm |
| For Use With | For HPLC, GC, TLC, SPE Application |
| Functionality/Interaction Type | Polar, Dipole-dipole, p-p interactions, hydrophobic |
| Hydrophobicity | Yes |
| Length (Metric) | 150 mm |
| Max. Pressure | 8700 psi (600 bar) |
| Mobile Phase Compatibility | Acetonitrile/water |
| Show More |