Learn More
Macherey-Nagel™ Nucleogel™ RP HPLC Guard Columns
Product Code. 12708052 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
3 mm
Unit Size:
Pack of 2
Product Code. 12708052
Brand: Macherey-Nagel™ 719542
Only null left
Specifications
pH 1 to 13
Particle Size 5 μm
Length (Metric) 5 mm
Pore Size 100Å
Type HPLC Guard Column
Diameter (Metric) Inner 3 mm
Stationary Phase NUCLEOGEL™ RP
Mobile Phase Compatibility Acetonitrile/Water
Solvents Acetonitrile/Water
Column Format Valco HPLC Guard Column
For Use With (Application) Separation of Small Molecules Such as Pharmaceuticals with Basic Properties
Technique HPLC
USP Type L21