missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleolin Antibody (364-5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47861CL1
This item is not returnable.
View return policy
Description
Nucleolin Monoclonal antibody specifically detects Nucleolin in Bovine (Negative),Human,Mouse (Negative),Rat (Negative) samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunofluorescence,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| Nucleolin | |
| Monoclonal | |
| CoraFluor 1 | |
| C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
| lysate of SU-DHL-1 Nuclei (Uniprot: P19338) | |
| 0.1 mL | |
| Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Stem Cell Markers | |
| 4691 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 364-5 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bovine (Negative), Human, Mouse (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction