missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleolin Antibody (NCL/7339), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20817C
This item is not returnable.
View return policy
Description
Nucleolin Monoclonal antibody specifically detects Nucleolin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Nucleolin | |
| Monoclonal | |
| DyLight 650 | |
| C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
| Recombinant full-length human Nucleolin protein | |
| 0.1 mL | |
| Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Stem Cell Markers | |
| 4691 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| NCL/7339 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction