missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleolin Antibody (NCL/8068R), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24068DL594
This item is not returnable.
View return policy
Description
Nucleolin Monoclonal antibody specifically detects Nucleolin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Nucleolin | |
| Monoclonal | |
| DyLight 594 | |
| C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
| Recombinant full-length human Nucleolin protein | |
| 0.1 mL | |
| Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Stem Cell Markers | |
| 4691 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| NCL/8068R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction