missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag DNA Bacteria for DNA purification from bacteria and yeast

Product Code. p-7193975
Click to view available options
No. of Reactions:
384 Preps
96 Preps
Unit Size:
384 reactions
96 reactions
This item is not returnable. View return policy

Product Code. 16475199

Brand: Macherey-Nagel™ 744310.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Magnetic bead based kit for the isolation of genomic DNA from bacteria and yeast.

  • Automation friendly solution for microbial samples
  • Environmentally sustainable buffer chemistry – no chaotropic salts
  • Combine with MN Bead Tubes for single sample processing or MN 96 Bead Plates for high throughput sample disruption
  • Liquid Proteinase K and Liquid RNase A for easy handling

Specifications

Capacity 0.4 μg/μL beads
Concentration Depending on amount and quality of sample
Content And Storage 18°C to 25°C
Isolation Technology Magnetic Bead
Sample Type Animal Tissue, Bacteria, Fungi, Human Tissue, Insects
Elution Volume 50 to 120 μL
For Use With (Application) Enzymatic Reactions, NGS, PCR, RT-PCR, Southern Blotting
No. of Reactions 96 Preps
Sample Volume <40 mg insects, <40 mg (108-109) bacteria, <30 mg fungi, <30 mg lipid rich tissue, <40 mg other tissues
Target DNA
Test Time 40 to 120 min/96 preps (excl. lysis)
Yield Depending on amount and quality of sample
Form Magnetic Beads
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.