missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoMag DNA/RNA Water kit for water and air samples
Click to view available options
No. of Reactions:
1 x 96
4 x 96
Unit Size:
384 reactions
96 reactions
Each
Pack of 4
Description
- Suitable for diverse salty and fresh water samples, ranging from turbid to clear as well as with air filters
- Minimized inhibition for reliable results
- Compatible with a variety of filters and filtration system
- Can be additionally combined with two different sizes of ceramic bead tubes for optimal sample lysis
- Manual and automated procedures at room temperature without reducing agents
- Typical downstream applications: qPCR, qRT-PCR, NGS
Specifications
Specifications
| Capacity | 0.4 μg/μL beads |
| Isolation Technology | Magnetic Bead |
| Sample Type | Air, Water |
| Efficiency | 0.4 μg/μL beads Binding capacity |
| No. of Reactions | 1 x 96 |
| Sample Volume | 300 bp - 50 kbp Fragments |
| Test Time | 40 min for KingFisher Flex (excl. Filtration and sample lysis) |
| Form | Highly Reactive Superparamagnetic Beads |
Safety and Handling
Product Identifier
- NucleoMag DNA/RNA Water (1x96)
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- MIXTURE LIST-Contains : guanidium thiocyanate, 2-propanol, guanidium hydrochloride,ethanol
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction