missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag Forensic for the purification of DNA from forensic samples

Product Code. 15819157
Change view
Click to view available options
:
Quantity: 4 x 96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15819157 Quantity: 4 x 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819157 Supplier Macherey-Nagel™ Supplier No. 744650.4

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

NucleoMag Forensic preps for the purification of DNA from forensic samples.

NucleoMag™ Forensic

  • One-tube procedure minimizes risk of cross-contamination
  • Small elution volumes > 25 μL possible
  • Easily adaptable for automated use
  • Forensic casework samples, contact traces (e.g., dried blood spots, cigarette filters, swabs)
  • Applications: STR analysis, qPCR, any kind of enzymatic reaction

Specifications

Concentration <1 ng/μL
Content And Storage Room temperature (18 - 25 °C)
Isolation Technology Magnetic Bead
Sample Type Casework Samples, Contact Traces
Efficiency Binding capacity: 0.4 μg/μL beads
Elution Volume Elution volume: 25 - 50 μL
For Use With (Equipment) Magnetic separator, separation plate
For Use With (Application) Isolation of genomic DNA from forensic samples
Includes NucleoMag F-Beads, buffers, Proteinase K
No. of Reactions 4 x 96 Preps
Test Time 40 - 120 min/96 preps (excl. lysis)
Yield Depends on sample amount and quality
Form Highly Reactive Superparamagnetic Beads
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.