Learn More
Cytiva Nucleon™ PhytoPure™ Genomic DNA Extraction Kits
Shop All Cytiva Products
Click to view available options
Quantity:
100 mg
1000 mg
Unit Size:
50 reactions
Description
Includes
Reagents 1 and 2 and PhytoPure Resin
- Rapid protocol does not require phenol or CTAB
- Two kit sizes provide economical scale-up using the same phenol-free protocol
- Effective removal of polysaccharides minimises inhibition of downstream applications
Specifications
Specifications
| Description | Quality, length, and purity of a phenol-chloroform extraction without phenol. Nucleon PhytoPure enables rapid chloroform extraction of high-quality, high molecular weight genomic DNA from plant and fungal samples with efficient removal of protein as well |
| Sample Type | Barley (Fresh, Frozen or Freeze Dried), Maize (Fresh, Frozen or Freeze Dried), Rye (Fresh, Frozen or Freeze Dried), Wheat (Fresh, Frozen or Freeze Dried) |
| For Use With (Application) | Successfully extracts DNA from many plant species, starting with fresh, frozen, or freeze dried material |
| Includes | Reagents 1 and 2 and PhytoPure™ Resin |
| Product Type | Nucleon PhytoPure |
| Quantity | 1000 mg |