missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleoporin 107 Antibody [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18854JF525
This item is not returnable.
View return policy
Description
Nucleoporin 107 Polyclonal antibody specifically detects Nucleoporin 107 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Nucleoporin 107 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Janelia Fluor 525 | |
| nuclear pore complex protein Nup107, nucleoporin 107kDa, NUP84Nucleoporin Nup107,107 kDa nucleoporin | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Nucleoporin 107 | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 57122 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction