missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleoporin 107 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19602
This item is not returnable.
View return policy
Description
Nucleoporin 107 Polyclonal specifically detects Nucleoporin 107 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Nucleoporin 107 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| nuclear pore complex protein Nup107, nucleoporin 107kDa, NUP84Nucleoporin Nup107,107 kDa nucleoporin | |
| Rabbit | |
| 106 kDa | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 57122 | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.44mg/mL | |
| Western Blot 1:500-1:3000, Immunoprecipitation 1:100-1:500 | |
| P57740 | |
| NUP107 | |
| Recombinant protein encompassing a sequence within the center region of human NUP107. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Species reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction