missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Nucleoredoxin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29531
This item is not returnable.
View return policy
Description
Nucleoredoxin Polyclonal antibody specifically detects Nucleoredoxin in Human,Mouse samples. It is validated for Western Blot
Specifications
| Nucleoredoxin | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 64359 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| EC 1.8.1.8, NRXFLJ12614, nucleoredoxin, nucleoredoxin 1, TRG-4 | |
| Between 385 and 435 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction