missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Nucleoside-Analog-1

Product Code. 16410678
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16410678 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16410678 Supplier MedChemExpress Supplier No. HY77651

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nucleoside-Analog-1 is a 4′-Azidocytidine analogue against Hepatitis C virus replication.

Specifications

Molecular Weight (g/mol) 267.2 g/mol
Product Type Reference compound
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Purity 95.0%
Quantity 5 mg
Solubility Information H2O : 1.89 mg/mL (7.07 mM∣Need ultrasonic)
For Use With (Application) COVID-19-anti-virus
CAS 876707-99-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.