missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Nucleoside-Analog-1
Specifications
Specifications
| Molecular Weight (g/mol) | 267.2 g/mol |
| Product Type | Reference compound |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Purity | 95.0% |
| Quantity | 100 mg |
| Solubility Information | H2O : 1.89 mg/mL (7.07 mM∣Need ultrasonic) |
| For Use With (Application) | COVID-19-anti-virus |
| CAS | 876707-99-2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?