Learn More
Macherey-Nagel™ Nucleosil™ 100-5 CN HPLC CC Guard Columns
Product Code. 12728122 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
4 mm
Unit Size:
Pack of 3
Product Code. 12728122
Brand: Macherey-Nagel™ 721604.40
Only null left
Specifications
Carbon Load 5%
pH 2 to 8
Particle Size 5 μm
Length (Metric) 8 mm
Pore Size 100Å
Type HPLC CC Guard Column
Diameter (Metric) Inner 4 mm
Stationary Phase NUCLEOSIL™ CN
Mobile Phase Compatibility n-heptane
Solvents n-heptane
Packing Material Silica
Crosslinking Cyano
Column Format Guard Column
For Use With (Application) HPLC
Technique HPLC
Particle Shape Spherical
Hydrophobicity Yes
Functionality/Interaction Type Hydrophobic, p-p
USP Type L10