Learn More
Macherey-Nagel™ Nucleosil™ 100-5 NH2-RP HPLC CC Guard Columns
Product Code. 10214002 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
4 mm
Unit Size:
Each
Product Code. 10214002
Brand: Macherey-Nagel™ 721605.40RP
Only null left
Specifications
Carbon Load 3.5%
pH 2 to 8
Particle Size 5 μm
Length (Metric) 8 mm
Pore Size 100Å
Type HPLC CC Guard Column
Diameter (Metric) Inner 4 mm
Stationary Phase NUCLEOSIL™ NH2-RP
Mobile Phase Compatibility Acetonitrile/Water
Solvents Acetonitrile/Water
Packing Material Silica
Crosslinking Amino
Column Format Guard Column
For Use With (Application) HPLC
Technique HPLC
Particle Shape Spherical
Hydrophobicity Yes
Functionality/Interaction Type Hydrophobic, Weak Ion Exchange
USP Type L8